Orders LinkContactUsLinkComponent
AdvanTip™ Pipette Tips
AdvanTip™ Pipette Tips
Low retention, 2 - 300 µl, -, 250, 300 μl pipettes, Bulk, 1000 tips/bag
Catalog # HAMI68009-20
Supplier:   HAMILTON BONADUZ
AdvanTip™ Pipette Tips
Catalog # HAMI68009-20
Supplier:  HAMILTON BONADUZ
AdvanTip™ Pipette Tips
AdvanTip™ Pipette Tips
Low retention, 2 - 300 µl, -, 250, 300 μl pipettes, Bulk, 1000 tips/bag
Price
99,50 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
  • Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
  • Certified free from RNase, DNase, and pyrogens
  • Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
  • Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
  • Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
Specifications
  • Packed:
    Bulk, 1000 tips/bag
  • Sterile:
    -
  • Volume:
    2 - 300 µl
  • Seq:
    03
  • Tip style:
    Low retention
  • For use with:
    250, 300 μl pipettes
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.