Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®

Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®

Glass bottle, amber, narrow mouth, 200 mL
Catalog # 4600-04
Supplier:   Avantor
Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®
Catalog # 4600-04
Supplier:  Avantor
CAS Number:  9005-64-5
Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®
Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®

Glass bottle, amber, narrow mouth, 200 mL

CAS Number:  9005-64-5
Price
2 034,50 €
Log in for availability
1 * 200 mL
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Density:1.08 kg (1 L)
  • Flash Pt:148.00 °C
Specifications

Specification Test Results

  • GMP Manufactured Product:
    ≤2.0
  • Meets E.P. Chemical Specifications:
    ≤0.25%
  • Meets B.P. Chemical Specifications:
    ≤10 ppm
  • Meets JPE Specifications:
    ≤1 ppm
  • Meets N.F. Requirements:
    ≤10 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    96 - 108
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Acid Value:
    ≤2.0
  • NF - Residue on Ignition:
    ≤0.25 %
  • NF - Dioxane:
    ≤10 ppm
  • NF - Ethylene Oxide:
    ≤1 ppm
  • NF - Hydroxyl Value, mg KOH/g:
    96 - 108
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Saponification Value, mg KOH/g:
    40 - 50
  • NF - Caproic Acid (Composition of Fatty Acids):
    ≤1.0 %
  • NF - Caprylic Acid (Composition of Fatty Acids):
    ≤10.0 %
  • NF - Capric Acid (Composition of Fatty Acids):
    ≤10.0 %
  • NF - Lauric Acid (Composition of Fatty Acids):
    40.0 - 60.0 %
  • NF - Myristic Acid (Composition of Fatty Acids):
    14.0 - 25.0 %
  • NF - Palmitic Acid (Composition of Fatty Acids):
    7.0 - 15.0 %
  • NF - Stearic Acid (Composition of Fatty Acids):
    ≤11.0 %
  • NF - Oleic Acid (Composition of Fatty Acids):
    ≤11.0 %
  • NF - Linoleic Acid (Composition of Fatty Acids):
    ≤3.0 %
  • NF - Peroxide Value:
    ≤5.0
  • NF - Water (H₂O):
    ≤3.0 %
  • EP/BP - Acid Value:
    ≤2.0
  • EP/BP - Total Ash:
    ≤0.25 %
  • EP/BP - Hydroxyl Value:
    96 - 108
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification D:
    Passes Test
  • EP/BP - Saponification Value:
    40 - 50
  • EP/BP - Ethylene Oxide:
    ≤1 ppm
  • EP/BP - Dioxan:
    ≤10 ppm
  • EP/BP - Water (H₂O):
    ≤3.0 %
  • EP/BP - Composition of Fatty Acids - Caproic Acid:
    ≤1.0 %
  • EP/BP - Composition of Fatty Acids - Caprylic Acid:
    ≤10.0 %
  • EP/BP - Composition of Fatty Acids - Capric Acid:
    ≤10.0 %
  • EP/BP - Composition of Fatty Acids - Lauric Acid:
    40.0 - 60.0
  • EP/BP - Composition of Fatty Acids - Myristic Acid:
    14.0 - 25.0 %
  • EP/BP - Composition of Fatty Acids - Palmitic Acid:
    7.0 - 15.0
  • EP/BP - Composition of Fatty Acids - Stearic Acid:
    ≤7.0 %
  • EP/BP - Composition of Fatty Acids - Oleic Acid:
    ≤11.0 %
  • EP/BP - Composition of Fatty Acids - Linoleic Acid:
    ≤3.0 %
  • EP - Peroxide Value:
    ≤10.0
  • JPE - Identification 1:
    Passes Test
  • JPE - Identification 2:
    Passes Test
  • JPE - Identification 3:
    Passes Test
  • JPE - pH at 25°C (5% Aqueous):
    4.0 - 7.0
  • JPE - Water (H₂O):
    ≤3.0 %
  • JPE - Acid Value:
    ≤4.0
  • JPE - Saponification Value:
    43 - 55
  • JPE - pH:
    4.0 - 7.0
  • JPE - Residue on Ignition:
    ≤0.25 %
  • JPE - Specific Gravity at 20°C:
    1.090 - 1.130
  • JPE - Viscosity (mm²/S):
    350 - 550
  • JPE - Heavy Metals (as Pb):
    ≤20 ppm
  • JPE - Arsenic (As):
    ≤2 ppm
  • Microbiological - Total Aerobic Microbial Count (cfu/g):
    ≤100
  • Microbiological - Escherichia coli:
    Passes Test
  • Microbiological - Pseudomonas aeruginosa:
    Passes Test
  • Microbiological - Salmonella:
    Passes Test
  • Microbiological - Staphylococcus aureus:
    Passes Test
  • Microbiological - Total Yeast and Mold Count (cfu/g):
    ≤50
  • Endotoxin Concentration (EU/g):
    ≤10
  • Color (APHA):
    ≤150
  • Formaldehyde:
    ≤10 ppm
  • Potassium (K):
    ≤5 ppm
  • Sodium (Na):
    ≤5 ppm
  • Ethylene glycol:
    ≤ 0.10 %
  • Diethylene glycol:
    ≤ 0.10 %

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    200 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.