Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions

pH buffer standard solution with certificate of analysis, 250 ml, 9,21, 250 mL
Catalog # HACHLZW9465.99
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9465.99
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 250 ml, 9,21, 250 mL

CAS Number:  
Price
20,30 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH buffer standard solution with certificate of analysis, 250 ml
  • pH value:
    9,21

Specifications