Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Labels, pipe marking

Pipe marking labels, Polyester, White/green, 150×12 mm, 5 labels/card
Catalog # BRDYN003298
Supplier:   Brady
Labels, pipe marking
Catalog # BRDYN003298
Supplier:  Brady
Labels, pipe marking
Labels, pipe marking

Pipe marking labels, Polyester, White/green, 150×12 mm, 5 labels/card

Price
12,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.

  • Promotes safety and simplifies maintenance
  • Arrows are pre-cut, and hence does not requires scissors additionally
  • Resistant to temperatures from –40 to +120 °C
  • Resistant to ultraviolet rays and conditions

The markers have a frame that contrasts with the color of the text to increase visibility.

Specifications
  • Colour:
    White/green
  • Description:
    Pipe marking labels
  • Dimensions:
    150×12 mm
  • Material:
    Polyester
  • Seq:
    11
  • Packed:
    5 labels/card
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.