Orders LinkContactUsLinkComponent
Accessories
Accessories
Filter fine, PG 8536
Catalog # MIEL1215141
Supplier:   MIELE
Accessories
Catalog # MIEL1215141
Supplier:  MIELE
Accessories
Accessories
Filter fine, PG 8536
Price
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Spécifications
  • Description:
    Filter fine
  • Seq:
    15
  • For:
    PG 8536