Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Ejection Rods

Sample Ejection Rods

Sample ejection rod, Stainless steel 316L, 700 mm
Catalog # SAMP8570A-700
Supplier:   Sampling Systems
Sample Ejection Rods
Catalog # SAMP8570A-700
Supplier:  Sampling Systems
Sample Ejection Rods
Sample Ejection Rods

Sample ejection rod, Stainless steel 316L, 700 mm

Price
208,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sample ejection rods are designed to facilitate the removal of samples.

  • A useful tool for removing samples
  • Choose a rod longer than the sampler being used
  • Available from stock
  • Fast delivery

The sample ejection rod is a convenient tool for removing any hard-to-eject samples collected within a sampler. Simply push the rod down the sampler shaft to eject the material.

Specifications
  • Description:
    Sample ejection rod
  • Length:
    700 mm
  • Material:
    Stainless steel 316L
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.