Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, solid sample holder (δ 1,5 - 4 mm, one-position) (B), for double beam only, sample size 120L×100W mm
Catalog # 634-6052
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-6052
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, solid sample holder (δ 1,5 - 4 mm, one-position) (B), for double beam only, sample size 120L×100W mm
Price
412,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cell holder, solid sample holder (δ 1,5 - 4 mm, one-position) (B), for double beam only, sample size 120L×100W mm
  • Seq:
    24