Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Four-cell holder for up to 100 mm (10 - 100 mm) square cuvette (B) for double beam only
Catalog # 634-6046
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-6046
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Four-cell holder for up to 100 mm (10 - 100 mm) square cuvette (B) for double beam only
Price
534,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Four-cell holder for up to 100 mm (10 - 100 mm) square cuvette (B) for double beam only
  • Seq:
    15