HIGH SALT WASH HBC BUFFER 1 * 160 mL
Catalog # PD099
Supplier: OMEGA BIO-TEK
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microscope Slides
Supplier: VWR Collection
DNA mini columns for genomic/plasmid DNA extraction, Hi-Bind®
Supplier: OMEGA BIO-TEK
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF