Order Entry
Belgium
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

500 Tests
Catalog # 13-6990-T500
Supplier:   Tonbo Biosciences
Propidium Iodide
Catalog # 13-6990-T500
Supplier:  Tonbo Biosciences
CAS Number:  25535-16-4
Propidium Iodide
Propidium Iodide

500 Tests

CAS Number:  25535-16-4
Price
65,80 €
Log in for availability
1 * 500 Tests
Product Details & Documents

Propidium Iodide (PI) is a membrane-impermeant DNA binding dye that cannot penetrate viable cells. PI rapidly enters cells with compromised membranes and intercalates between base pairs allowing exclusion of non-viable cells from analysis of flow cytometry data. PI has a broad emission spectrum from 535-617 nm and can be detected in either FL2 or FL3 detectors. When used with Annexin V FITC, it is recommended to analyze PI in FL2.

  • Formulation: 10 mM NaH₂PO₄, 150 mM NaCl, 0,09% NaN₃, pH 7,2
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
Specifications

Specifications

  • Pk:
    500 Tests

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.