Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

General purpose sample storage vials and caps

22,00 ml, 23×85 mm, Amber, Yes, 20-400, PP, PTFE/silicone
Catalog # HEWL5183-4323
Supplier:   Agilent
General purpose sample storage vials and caps
Catalog # HEWL5183-4323
Supplier:  Agilent
General purpose sample storage vials and caps
General purpose sample storage vials and caps

22,00 ml, 23×85 mm, Amber, Yes, 20-400, PP, PTFE/silicone

Price
128,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Specifications
  • Cap included:
    Yes
  • Cap material:
    PP
  • Cap Size:
    20-400
  • Ø×H:
    23×85 mm
  • Septum:
    PTFE/silicone
  • Capacity:
    22,00 ml
  • Colour:
    Amber
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.