ContactUsLinkComponent
DB-225MS 15M 0.25MM 0.25UM COL TOROID 1 * 1 items
  HEWL222-2912LTM
 :  Agilent
DB-225MS 15M 0.25MM 0.25UM COL TOROID 1 * 1 items
  HEWL222-2912LTM
 :  Agilent
Frequently Bought Together