Order Entry
Belgium
Orders LinkContactUsLinkComponent
Capillary columns, DB-624

Capillary columns, DB-624

7890/6890 LTM module, Capillary, DB-624, 1,12 µm, 0,20 mm, 10 m
Catalog # HEWL128-1314LTM
Supplier:   Agilent
Capillary columns, DB-624
Catalog # HEWL128-1314LTM
Supplier:  Agilent
Capillary columns, DB-624
Capillary columns, DB-624

7890/6890 LTM module, Capillary, DB-624, 1,12 µm, 0,20 mm, 10 m

Price
1 620,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.

  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    1,12 µm
  • Ø int.:
    0,20 mm
  • Description:
    7890/6890 LTM module
  • Length:
    10 m
  • Phase:
    DB-624