Orders LinkContactUsLinkComponent
Operating supplies
Operating supplies
Operating supplies set, SIPS 10/20
Catalog # VARI190025400
Supplier:   VARIAN
Operating supplies
Catalog # VARI190025400
Supplier:  VARIAN
Operating supplies
Operating supplies
Operating supplies set, SIPS 10/20
Price
1 850,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 L), a three-way SIPS tee piece assembly for SIPS-20, six SIPS pump tubings, a SIPS kit and ten pack SIPS pump bands.
Specifications
  • Model:
    SIPS 10/20
  • Description:
    Operating supplies set
  • Seq:
    01