32p ph meter pkg w/triode electrode contains:meter,electrode holder triode electrode,atc probe and ph buffers 1 * 1 items
HEWLG4392A
: Agilent
32p ph meter pkg w/triode electrode contains:meter,electrode holder triode electrode,atc probe and ph buffers 1 * 1 items
HEWLG4392A
: Agilent
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK