Order Entry
Belgium
Orders LinkContactUsLinkComponent
HPLC columns, ZORBAX® Eclipse XDB

HPLC columns, ZORBAX® Eclipse XDB

Method Validation Kit, Eclipse XDB-C18, —, 5 µm, 4,60 mm, 150 mm
  HEWL993967-902K
 :   Agilent
HPLC columns, ZORBAX® Eclipse XDB
  HEWL993967-902K
 :  Agilent
HPLC columns, ZORBAX® Eclipse XDB
HPLC columns, ZORBAX® Eclipse XDB

Method Validation Kit, Eclipse XDB-C18, —, 5 µm, 4,60 mm, 150 mm

 
2 420,00 €
   
1 * 1 ST
 

Good peak shape for basic, acidic and neutral compounds.

  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping
 
  • Column Format:
    Method Validation Kit
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
  • Length:
    150 mm
  • Seq:
    102
  • Phase:
    Eclipse XDB-C18
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.