ContactUsLinkComponent
Variable angle sample holder, jaw syle centre mount, dra-ca-3 for cary 1/3 allows samples to be positioned at center of dra for meas urement. suitable for rigid, opaque samples e.g. mirrors 1 * 1 items
Catalog # VARI7910037300
Supplier:  VARIAN
Variable angle sample holder, jaw syle centre mount, dra-ca-3 for cary 1/3 allows samples to be positioned at center of dra for meas urement. suitable for rigid, opaque samples e.g. mirrors 1 * 1 items
Catalog # VARI7910037300
Supplier:  VARIAN
Frequently Bought Together