Inlet liners
Liner, ultra inert, 2 mm, dimpled, splitless, 2 mm
N° de catalogue HEWL5190-2297
Fournisseur: Agilent
Aller à :
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Spécifications
- Ø int.:2 mm
- Description:Liner, ultra inert, 2 mm, dimpled, splitless
- Seq:11