Threaded ball flanges
Socket KF 25 with steel thread M 30×1,5
Catalog # LENZ05108230
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Made of stainless steel.
- Very good chemical resistance
- High temperature resistance
- Sturdy
With KF socket, with thread.
Specifications
- Type:Socket KF 25 with steel thread M 30×1,5