Acetone, J.T.Baker®
10 L
Supplier: Avantor
Acetone, J.T.Baker®
Catalog # 3413.9010
Supplier: Avantor
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- UN:1090
- ADR:3,II
Documents
Specifications
Specification Test Results
- Assay (by GC):min. 99 %
Specifications
- Pk:10 L
- Cat. No.:3413.9010
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



