Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, J.T.Baker®

Acetone, J.T.Baker®

5 L
Catalog # 3413.5000
   Safer Human and Environmental Health
Supplier:   Avantor
Acetone, J.T.Baker®
Catalog # 3413.5000
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone, J.T.Baker®
Acetone, J.T.Baker®

5 L

CAS Number:  67-64-1
Price
Inquire For Price
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • UN:1090
  • ADR:3,II
Specifications

Specification Test Results

  • Assay (by GC):
    min. 99 %

Specifications

  • Pk:
    5 L
  • Cat. No.:
    3413.5000
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.