ContactUsLinkComponent
C18G x 3µm particle size x 200Å pore size, 100mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C181-03M03P
C18G x 3µm particle size x 200Å pore size, 100mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C181-03M03P
Frequently Bought Together