ContactUsLinkComponent
HILIC FL x 5µm particle size x 120Å pore size, 250mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL8-05R46S
undefined
HILIC FL x 5µm particle size x 120Å pore size, 250mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL8-05R46S
Frequently Bought Together