ContactUsLinkComponent
HILIC RP x 5µm particle size x 120Å pore size, 150mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-05N03P
undefined
HILIC RP x 5µm particle size x 120Å pore size, 150mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-05N03P
Frequently Bought Together