ContactUsLinkComponent
HILIC RP x 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-03N46P
undefined
HILIC RP x 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-03N46P
Frequently Bought Together