ContactUsLinkComponent
Cyano x 5µm particle size x 120Å pore size, 250mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05R02P
Cyano x 5µm particle size x 120Å pore size, 250mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05R02P
Frequently Bought Together