ContactUsLinkComponent
C8H x 3µm particle size x 1000Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C82-03L02S
C8H x 3µm particle size x 1000Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C82-03L02S
Frequently Bought Together