ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 50mm length x 300µm ID, PEEKsil with guard ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03L30K
C18G x 3µm particle size x 120Å pore size, 50mm length x 300µm ID, PEEKsil with guard ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03L30K
Frequently Bought Together