Orders LinkContactUsLinkComponent
Connection system GL 45
Connection system GL 45
Screw cap, GL 45, PP, with three ports GL 14
Catalog # BIBB4537/04
Supplier:   SciLabware
Connection system GL 45
Catalog # BIBB4537/04
Supplier:  SciLabware
Connection system GL 45
Connection system GL 45
Screw cap, GL 45, PP, with three ports GL 14
Price
86,70 €
Log in for availability
1 * 2 ST
Product Details & Documents
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
  • Temperature resistant to 140 °C
  • Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
  • Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membrane filter with 0,2 μm sized pores permits sterile pressure equalisation. Unused ports can be sealed with a blanking cap. Components are fully interchangeable and compatible with both two and three port screwcaps.
Specifications
  • Description:
    Screw cap, GL 45, PP, with three ports GL 14
  • Seq:
    02