Orders LinkContactUsLinkComponent
Stopcocks, Galtek®
Stopcocks, Galtek®
Inlet 1/4" integral ferrule, outlet 1/4" integral ferrule, length 72,9 mm
Catalog # ENTEPVT4-T4-2
Supplier:   ENTEGRIS
Stopcocks, Galtek®
Catalog # ENTEPVT4-T4-2
Supplier:  ENTEGRIS
Stopcocks, Galtek®
Inlet 1/4" integral ferrule, outlet 1/4" integral ferrule, length 72,9 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Two- or three way, ¹/₈" orifice.
  • All wetted parts are PFA and PTFE.
  • Small compact design provides minimal internal volume.
  • Several connection sizes available to choose from
Specifications
  • Description:
    Inlet 1/4" integral ferrule, outlet 1/4" integral ferrule, length 72,9 mm