Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Stopcocks, Galtek®
Stopcocks, Galtek®
Inlet 1/4" MNPT, outlet 1/4" MNPT, length 43,2 mm
Catalog # ENTEPV4-4-2
Supplier:   ENTEGRIS
Stopcocks, Galtek®
Catalog # ENTEPV4-4-2
Supplier:  ENTEGRIS
Stopcocks, Galtek®
Inlet 1/4" MNPT, outlet 1/4" MNPT, length 43,2 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Two- or three way, ¹/₈" orifice.
  • All wetted parts are PFA and PTFE.
  • Small compact design provides minimal internal volume.
  • Several connection sizes available to choose from
Specifications
  • Désignation:
    Inlet 1/4" MNPT, outlet 1/4" MNPT, length 43,2 mm