ContactUsLinkComponent
Poly tubing #LT106K4800 150µ Material: low density polyethylene (LDPE) Alkenyl CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 48´ (1219mm) Packing: Roll of 500 '(150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K4800
Supplier:  KNF CLEANROOM
Poly tubing #LT106K4800 150µ Material: low density polyethylene (LDPE) Alkenyl CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 48´ (1219mm) Packing: Roll of 500 '(150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K4800
Supplier:  KNF CLEANROOM
Frequently Bought Together