Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps

Forceps

Straight, rounded tip (1,5 mm radius, 3,0 mm thickness), 178 mm
Catalog # ENTEC16-0315
Supplier:   ENTEGRIS
Forceps
Catalog # ENTEC16-0315
Supplier:  ENTEGRIS
Forceps

Straight, rounded tip (1,5 mm radius, 3,0 mm thickness), 178 mm

Price
Demander un prix
Connexion Pour la disponibilité
1 * 4 ST
Product Details & Documents

ETFE, white. Durable forceps for substrate handling.

  • Different tip styles available
  • Packaged in protective cases
Spécifications
  • Type:
    Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
  • Length:
    178 mm