ContactUsLinkComponent
Poly tubing #LT106K:3600 150µ Material: low density polyethylene (LDPE) Alkenyl CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 36 ´(914mm) Packing: Roll of 500 '(150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K3600
Supplier:  KNF CLEANROOM
Poly tubing #LT106K:3600 150µ Material: low density polyethylene (LDPE) Alkenyl CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 36 ´(914mm) Packing: Roll of 500 '(150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K3600
Supplier:  KNF CLEANROOM