Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps

Forceps

Straight, rounded tip (1,5 mm radius, 3,0 mm thickness), 124 mm
Catalog # ENTEC12-0315
Supplier:   ENTEGRIS
Forceps
Catalog # ENTEC12-0315
Supplier:  ENTEGRIS
Forceps

Straight, rounded tip (1,5 mm radius, 3,0 mm thickness), 124 mm

Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents

ETFE, white. Durable forceps for substrate handling.

  • Different tip styles available
  • Packaged in protective cases
Specifications
  • Type:
    Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
  • Length:
    124 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.