Sample dipper with v bottom
Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm
Catalog # ENTED15-0215
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Holds a wafer or a square plate.
- Slot has 'V' bottom
- Clip on handle slides down to hold substrates in place
- Hold one wafer or one square plate
Specifications
- Description:Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm