Forceps
Straight, rounded tip (0,8 mm radius), 127 mm
ENTEC01-0315
: ENTEGRIS
ETFE, white. Durable forceps for substrate handling.
- Different tip styles available
- Packaged in protective cases
- Type:Straight, rounded tip (0,8 mm radius)
- Length:127 mm



