Sample dipper with v bottom
Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm
ENTED14-0215
: ENTEGRIS
Holds a wafer or a square plate.
- Slot has 'V' bottom
- Clip on handle slides down to hold substrates in place
- Hold one wafer or one square plate
- Description:Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm