Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bouffant caps, PP

Bouffant caps, PP

Blue, 530 mm
Catalog # 113-9219
Bouffant caps, PP
Catalog # 113-9219
Bouffant caps, PP
Bouffant caps, PP

Blue, 530 mm

Price
168,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents

Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.

  • Single elastic band
  • Latex-free
  • Soft and comfortable

Weight: 10 g/m²

Packaging: 100 pieces per polybag. 20 bags per carton.

Specifications
  • Ø:
    530 mm
  • Colour:
    Blue
  • Seq:
    03
  • Cat. No.:
    113-9219
  • Garment application:
    Medical|Clinical