Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Beard cover, PP

White, 220×450 mm, 100/bag
  114-9126
Beard cover, PP
  114-9126
Beard cover, PP
Beard cover, PP

White, 220×450 mm, 100/bag

 
87,10 €
   
1 * 1.000 ST
 

This disposable beard cover is made from soft non woven PP.

  • Single elastic headband
  • Soft, breathable and lightweight
  • Latex-free elasticated edges and headband to reduce the risk of allergic reactions

Weight: 10 to 12 g/m²

 
  • Packed:
    100/bag
  • Colour:
    White
  • Seq:
    01
  • W×L:
    220×450 mm
  • Cat. No.:
    114-9126
  • Weight:
    10 - 12 g/m²