Orders LinkContactUsLinkComponent
Sample dipper with v bottom
Sample dipper with v bottom
Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
Catalog # ENTED11-0215
Supplier:   ENTEGRIS
Sample dipper with v bottom
Catalog # ENTED11-0215
Supplier:  ENTEGRIS
Sample dipper with v bottom
Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents
Holds a wafer or a square plate.
  • Slot has 'V' bottom
  • Clip on handle slides down to hold substrates in place
  • Hold one wafer or one square plate
Documents
Specifications
  • Description:
    Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.