Bouffant caps, PP
Blue, 580 mm
Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.
- Single elastic band
- Latex-free
- Soft and comfortable
Weight: 16 to 23 g/m²
Packaging: 100 pieces per polybag. 2000 pieces per carton.
Documents
- Ø:580 mm
- Colour:Blue
- Seq:05
- Cat. No.:113-9225
- Garment application:Medical|Clinical



