Bouffant caps, PP
White, 610 mm
Catalog # 113-9226
Supplier: Hubei Orient International
Jump to:
Product Details & Documents
Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.
- Single elastic band
- Latex-free
- Soft and comfortable
Weight: 16 to 23 g/m²
: 100 pieces per polybag. 2000 pieces per carton.
Specifications
- Ø:610 mm
- Colour:White
- Seq:06
- Cat. No.:113-9226
- Garment application:Medical|Clinical
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



