Jars with screw cap
with polypropylene screw cap, labelled, irradiated, 150 ml
Catalog # 216-2048
Supplier: Anicrin
Jump to:
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
- Twist cap for quick opening and closing
- Very good temperature resistance
- Very good chemical resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Type:with polypropylene screw cap, labelled, irradiated
- Capacity:150 ml
- Seq:04