Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forskolin

Forskolin

Glass bottle for solids, 10 mg
Catalog # CAYM11018-10
Supplier:   Cayman Chemical
Forskolin
Catalog # CAYM11018-10
Supplier:  Cayman Chemical
CAS Number:  66575-29-9
Forskolin

Glass bottle for solids, 10 mg

CAS Number:  66575-29-9
Price
163,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
  • CAS Number:66575-29-9
  • Formula:C₂₂H₃₄O₇
  • MDL Number:MFCD00082317
  • Molecular Weight:410.51 g/mol
  • EINECS:266-410-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.