Forskolin
Glass bottle for solids, 5 mg
Catalog # CAYM11018-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- MDL Number:MFCD00082317
- Molecular Weight:410.51 g/mol
- EINECS:266-410-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg



