Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cycloheximide

Cycloheximide

Glass bottle for solids, 100 mg
Catalog # CAYM14126-100
Toxic
Health
Environment
Supplier:   Cayman Chemical
Cycloheximide
Catalog # CAYM14126-100
Supplier:  Cayman Chemical
CAS Number:  66-81-9
Cycloheximide
Cycloheximide

Glass bottle for solids, 100 mg

CAS Number:  66-81-9
Price
55,50 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.