Order Entry
Belgium
Orders LinkContactUsLinkComponent
Anisomycin

Anisomycin

Glass bottle for solids, 10 mg
Catalog # CAYM11308-10
Supplier:   Cayman Chemical
Anisomycin
Catalog # CAYM11308-10
Supplier:  Cayman Chemical
CAS Number:  22862-76-6
Anisomycin

Glass bottle for solids, 10 mg

CAS Number:  22862-76-6
Price
73,60 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
  • CAS Number:22862-76-6
  • Formula:C₁₄H₁₉NO₄
  • MDL Number:MFCD00077650
  • Molecular Weight:265.31 g/mol
  • EINECS:245-269-7
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications