Order Entry
Belgium
Orders LinkContactUsLinkComponent
Homogenisers, behrotest®

Homogenisers, behrotest®

Homogeniser, 20 litre capacity, QMR 20
Catalog # BEHRB00218333
Supplier:   BEHR
Homogenisers, behrotest®
Catalog # BEHRB00218333
Supplier:  BEHR
Homogenisers, behrotest®
Homogenisers, behrotest®

Homogeniser, 20 litre capacity, QMR 20

Price
4 740,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Homogenisers for pretreatment, sample splitting and homogenisation of heterogeneous water samples in accordance with German waste water quality control standards.

  • The sample vessel is protected against damage by the homogeniser frame, which has two handles for easy carrying
  • Sample vessel volumes from 5 to 25 litre permit multiple sample collection (mixed samples)
  • Graduated for easy measurement of sample volume
  • Adjustable-speed magnetic stirrer
  • Withdrawl of desired volume of homogenized sample by means of a PTFE stopcock
  • Can be operated from a 12 V auto battery using an optional voltage converter
Specifications
  • Description:
    Homogeniser, 20 litre capacity, QMR 20
  • Seq:
    03
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Behrotest glass vessel, 15 l
    Behrotest glass vessel, 15 l
    331-3312
    Log in for availability
    1 * 1 ST - 1 340,00 €

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.