Orders LinkContactUsLinkComponent
Digestion system, behrotest®
Digestion system, behrotest®
Hydrolysis unit, complete for one sample, HYDRO 1
Catalog # BEHRB00218430
Supplier:   BEHR
Digestion system, behrotest®
Catalog # BEHRB00218430
Supplier:  BEHR
Digestion system, behrotest®
Digestion system, behrotest®
Hydrolysis unit, complete for one sample, HYDRO 1
Price
611,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
Delivery information: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Specifications
  • Description:
    Hydrolysis unit, complete for one sample
  • Seq:
    01
  • Type:
    HYDRO 1